Anti-17-Hydroxypregnenolone Polyclonal Antibody
ABNOPAB5162
: Abnova
- Antibody type:Primary
- Antigen name:17-Hydroxypregnenolone
- Antigen symbol:17-Hydroxypregnenolone
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,5 (50% glycerol, 0,01% thimerosal)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:4-PREGNEN-17-OL-3, 20-DIONE 3-CARBOXYMETHYLOXIME.
- Size:150 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against 17-hydroxyprogesterone.
Type: Primary
Antigen: 17-Hydroxypregnenolone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: