Order Entry
Singapore
ContactUsLinkComponent
Anti-BSD Rabbit Polyclonal Antibody
Anti-BSD Rabbit Polyclonal Antibody
Catalog # ABNOPAB7931
Supplier:  Abnova
Anti-BSD Rabbit Polyclonal Antibody
Catalog # ABNOPAB7931
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Aspergillus Terrus Blasticidin S Deaminase
  • Antigen symbol:
    BSD
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Fungus
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (50% glycerol, 1 mg/mL BSA, 0,05% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide (conjugated with KLH) corresponding to N-terminus of Aspergillus Terrus Blasticidin S Deaminase.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of Aspergillus Terrus Blasticidin S Deaminase.

Recommended Dilutions: ELISA: 1:4000; Western Blot: 1:2000

Type: Primary
Antigen: BSD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Fungi