Anti-IgG Rabbit Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # ABNOPAB10714
Supplier: Abnova
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Monoclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Epitope:F(ab')2
- Cross adsorption:No
- Format:Lyophilized
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C on dry atmosphere. After reconstitution with 1 mL of deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Human IgG F(ab')2 fragment
- Purification:solid phase adsorption
- Size:2 mg
- Pk:2 mg
Specifications
About this item
Rabbit anti-human IgG recognizes human IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Rhodamine.
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: F(ab')2
Host: Rabbit
Isotype:
Reactivity: Human