Distillation system, cyanide, MIDI-VAP™ 4000, KIMBLE®
Catalog # KIML479400-4000
Supplier: KIMBLEKONTES
Specifications
- Description:Distillation system, cyanide complete, MIDI-VAP™ 4000
- Pk:1
Specifications
About this item
The Midi-Vap 4000™ distillation system is designed to support the semi-automatic colorimetry methods for drinking, ground, surface and saline waters as well as domestic and industrial wastes.
- Easy to read LED display
- Automatic shutoff at test completion
The system features a ten-position heater block for consistent, even heating and a Watlow EZ-Zone digital controller.
Certifications: Glassware conforms to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery: Includes ten sets of glassware used for the analysis of cyanide.