Microscale organic chemistry kit, threaded, MICROFLEX®, KIMBLE®
Catalog # KIML747000-0005
Supplier: KIMBLEKONTES
Specifications
- Description:Microscale standard threaded kit, in EKONO-CASE™
- Pk:1 KIT
Specifications
About this item
This chemistry kit is designed for most common organic chemistry experiments.
The whole components are packed in a rugged polyethylene storage case and also the kit fits easily into lab bench drawers. It has blue nylon connectors that are highly resistant to temperature upto 200 °C.
Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.