Order Entry
Singapore
ContactUsLinkComponent
Screw caps, open top, KIMBLE®
Screw caps, open top, KIMBLE®
Catalog # KIML410119-1305
Supplier:  KIMBLEKONTES
undefined
Screw caps, open top, KIMBLE®
Catalog # KIML410119-1305
Supplier:  KIMBLEKONTES
Specifications
  • Thread:
    13-425
  • Description:
    Screw cap, blue, with hole Ø 5,5 mm
  • Pk:
    12
Specifications
About this item
Gas filled nylon cap, with open top. This cap has modified GPI thread finish to accommodate us with an O-ring seal.
  • Temperature limit: 150 ºC