Order Entry
Singapore
ContactUsLinkComponent
Gas burettes, Tutwiler, KIMBLE®
Gas burettes, Tutwiler, KIMBLE®
Catalog # KIML30034A-100
Supplier:  KIMBLEKONTES
undefined
Gas burettes, Tutwiler, KIMBLE®
Catalog # KIML30034A-100
Supplier:  KIMBLEKONTES

Specifications

  • Capacity:
    100 ml
  • Length:
    480 mm
  • Pk:
    1
  • Tolerance:
    0,2 ± ml

Specifications

About this item

Borosilicate glass.

  • Designed for use with method UOP 9-85
  • Large inner diameter connections reduce clogging and deposits of the precipitates

These burettes designed for use with high sulphur content gas streams.

Delivery: Supplied with a size 4 glass stopcock in the bottom, a size 2 stopcock in the top and a size 13 glass pennyhead stopper.