Order Entry
Singapore
ContactUsLinkComponent
Centrifuge tubes, conical bottom, beaded rim, graduated, KIMBLE®
Centrifuge tubes, conical bottom, beaded rim, graduated, KIMBLE®
Catalog # KIML45167-50
Supplier:  KIMBLEKONTES
undefined
Centrifuge tubes, conical bottom, beaded rim, graduated, KIMBLE®
Catalog # KIML45167-50
Supplier:  KIMBLEKONTES

Specifications

  • Closure type:
    Without cap
  • Ø×L:
    30×133 mm
  • Capacity:
    50 ml
  • Pk:
    1

Specifications

About this item

Borosilicate glass, KIMAX®, graduated, beaded.

  • Calibrated to contain
  • Graduation in durable black ceramic enamel

Used in the determination of the solubility index of dry milk solids or where products as specified by the American Dairy Products Institute. Also used in pulp determination of citrus products.

Tubes are graduated upward to 1,0 ml in divisions of 0,1 ml; from 1 to 2 ml in divisions of 0,2 ml; from 2 to 10 ml in divisions of 0,5 ml; from 10 to 20 ml in divisions of 1 ml.

Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.