Order Entry
Singapore
ContactUsLinkComponent
Volumetric flasks, acc. to Kohlrausch, glass, clear, KIMAX®, KIMBLE®
  KIML28100-100
Volumetric flasks, acc. to Kohlrausch, glass, clear, KIMAX®, KIMBLE®
  KIML28100-100
  • Capacity:
    100 ml
  • Height:
    159 mm
  • Pk:
    12
  • Tolerance:
    ±0,10 ml
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Graduation ring is blasted on the neck
  • Calibrated to contain
  • With a marking spot
Flask is calibrated with an enlarged mouth that provides an easy introduction of samples of solid materials such as sugar beets.
 : Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.