Volumetric flasks, class A, serialised, glass stopper, KIMBLE®
Borosilicate glass KIMAX®, with glass stopper.
- Calibrated to contain (TC)
- Wide base for greater stability
- Graduation ring blasted on the neck
Flasks are ideal for measuring accurate volumes of liquids and have a mark. These flasks are permanently marked with an individual serial number and a marking spot.
Designed from ASTM specification E237. Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.