Order Entry
Singapore
ContactUsLinkComponent
Flasks, pear shaped, with standard ground joint, one neck, KIMBLE®
Flasks, pear shaped, with standard ground joint, one neck, KIMBLE®
Catalog # KIML294250-0050
Supplier:  KIMBLEKONTES
Flasks, pear shaped, with standard ground joint, one neck, KIMBLE®
Catalog # KIML294250-0050
Supplier:  KIMBLEKONTES
Specifications
  • Socket size:
    14/20 NS
  • Capacity:
    50 ml
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
This flask is with a standard taper joint at top and made of heavy wall.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.