Order Entry
Singapore
ContactUsLinkComponent
Pasteur pipettes, disposable, KIMBLE®
Pasteur pipettes, disposable, KIMBLE®
Catalog # KIML63B93P
Supplier:  KIMBLEKONTES
undefined
Pasteur pipettes, disposable, KIMBLE®
Catalog # KIML63B93P
Supplier:  KIMBLEKONTES

Specifications

  • Ext.Ø:
    7
  • Packed:
    1000/case
  • Capacity:
    2 ml
  • Description:
    Disposable pasteur pipettes, plugged
  • Length:
    228 mm
  • Seq:
    04
  • Pk:
    1.000

Specifications

About this item

Soda Lime glass, disposable.

  • Intended for one-time use in the transfer of small volumes
  • With or without cotton plug
  • Soda lime pipettes are manufactured from 90 expansion soda lime glass conforming to USP Type III requirements