Micro capillaries, disposable, KIMBLE®
Catalog # KIML71900-100
Supplier: KIMBLEKONTES
Specifications
- Description:Capillary pipettes, 100 µl, colour code blue
- Length:127 mm
- Seq:02
- Pk:250
Specifications
About this item
Soda lime glass pipettes, USP Type III.
- Calibrated to contain
- Colour-coded
- Marked with single black capacity ring
- Packaged in easy-open cylinders for convenient dispensing
Certifications: Designed from ASTM Specification E672.
Packaging: Each cylinder contains 250 pipettes and one pipette device.