Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microplates
PCR Plates
PCR plates
PCR plates
PCR plates
Catalog # BITX63300634
Supplier:
BIOTIX
Expand image
PCR plates
Catalog # BITX63300634
Supplier:
BIOTIX
Specifications
No. of wells:
96
Skirt:
Semi-skirted
Well colour:
Clear
Seq:
02
Pk:
100
Well volume:
200 μl
Specifications
About this item
Product Details
Product Details