Order Entry
Singapore
ContactUsLinkComponent
Flasks, round bottom, short neck, KIMBLE®
Flasks, round bottom, short neck, KIMBLE®
  KIML25285-250
Flasks, round bottom, short neck, KIMBLE®
  KIML25285-250

 

  • Ext.Ø:
    83 mm
  • Socket size:
    24/40 NS
  • Capacity:
    250 ml
  • Height:
    140 mm
  • Seq:
    01
  • Pk:
    12

 

 

Manufactured from 33 expansion, low extractable borosilicate glass, KIMAX®.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

This round bottom boiling flask is with a short neck and a medium length standard taper joint.

 : Designed from ASTM specification E1403, Type II, Class II requirements. Glass conforming to to USP Type I and ASTM E438, Type I, Class A requirements.