Order Entry
Singapore
ContactUsLinkComponent
Filtering flasks, ULTRA-WARE®, KimCote®, KIMBLE®
  KIML953760-1002
Filtering flasks, ULTRA-WARE®, KimCote®, KIMBLE®
  KIML953760-1002
  • Capacity:
    1000 ml
  • Pk:
    1
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
These erlenmeyer flasks are heavy walled to ensure the mechanical strength needed for vacuum filtration and the safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals. All flasks have a side arm for connection to 6 mm int.Ø flexible tubing.
 : Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.