Order Entry
Singapore
Orders LinkContactUsLinkComponent
Volumetric flasks, class A, KIMBLE®
Volumetric flasks, class A, KIMBLE®
Supplier:   KIMBLEKONTES
Volumetric flasks, class A, KIMBLE®
Supplier:  KIMBLEKONTES
Certificates
Volumetric flasks, class A, KIMBLE®
2 Options Available Below
Product Details & Documents
Borosilicate glass, KIMAX® with PTFE/rubber-lined phenolic cap.
  • Single graduation ring blasted on the neck
  • Calibrated to contain
  • Screw threaded flask
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Product Family Options
Product Information
  • Capacity
    Thread
    Height
    Tolerance
    Availability
    Price
  • 5 ml
    13-425
    80 mm
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $199.00
    $199.00
    1 * 1 items
       $199.00
  • 10 ml
    13-425
    90 mm
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $259.00
    $259.00
    1 * 1 items
       $259.00