Measuring cylinders, tall form, class A, borosilicate glass, educational grade, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
Certificates
About this item
These cylinders, graduated, manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements, are ideal for use in educational institutions and for many general laboratory procedures.
- Calibrated to deliver (TD)
- Pour spout
- Hexagonal plastic base
- Graduations in durable blue ceramic enamel
- 10 ml cylinder has a funnel shaped top
Delivery: Supplied with SAFE-GARD® bumpers.