Order Entry
Singapore
ContactUsLinkComponent
Test Tubes, with stopper, KIMAX®, KIMBLE
  KIML45100-25200
Test Tubes, with stopper, KIMAX®, KIMBLE
  KIML45100-25200
  • Ext.Ø:
    25 mm
  • Capacity:
    65 ml
  • Length:
    200 mm
  • Pk:
    24
Borosilicate glass, with pennyhead stoppers, KIMAX®.
  • Durable white ceramic enamel marking spot on each tube
These test tubes have round bottom with uniform wall thickness.
 : Designed from ASTM E982, type III.
Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.