Order Entry
Singapore
ContactUsLinkComponent
Cleanroom wipes, pre-saturated, sterile, KIMTECH PURE* W4
  KIMB76490
undefined
Cleanroom wipes, pre-saturated, sterile, KIMTECH PURE* W4
  KIMB76490
  • Packed:
    40 wipes/pouch, 5 pouches/bag, 2 bags/case
  • Colour:
    White
  • L×W:
    279×229 mm
  • Pk:
    400
This sterile non woven PP wipe is pre-saturated with 70% isopropyl alcohol and 30% deionized water. Recommended for the most critical wiping tasks in ISO Class 4 or higher cleanroom environments.
  • Ideal one-step application of alcohol to critical surfaces
  • Resealable pouch with case liner
Suitable for use in class 10, ISO 4, M2.5 cleanrooms.