Anti-ADAM12 Goat Polyclonal Antibody
PIEAPA518314
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase Domain 12
- Antigen symbol:ADAM12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:8038
- Antigen synonyms:a disintegrin and metalloproteinase domain 12 (meltrin alpha)|ADAM metallopeptidase domain 12|mouse|homolog of)|ADAM12|MLTNA|RP11-295J3.5|meltrin alpha|A disintegrin and metalloproteinase domain 12 (Meltrin-alpha|MLTN|MCMP|MCMPMltna
- Storage buffer:TBS, 0.5% BSA, 0.02% sodium azide, pH 7.3
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide sequence (AARPLPVSPARALC) corresponding to the N-terminus amino acids of ADAM12
- Purification:Antigen affinity chromatography
- Size:100 ug
- Pk:100 µG
This gene encodes a member of the ADAM protein family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene has two alternatively spliced transcripts: a shorter secreted form and a longer membrane-bound form. The shorter form is found to stimulate myogenesis.
Immunohistochemistry: 3-5µg/ml, Western Blot: 0.1-0.3 ug/ml
Type: Primary
Antigen: ADAM12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human