Anti-Mitofilin Rabbit Polyclonal Antibody
Catalog # PIEAPA3870
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Primary
- Antigen name:Mitofilin
- Antigen symbol:Mitofilin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse,Cat
- Western blot:Yes
- Environmentally Preferable:
- Format:whole serum
- Form:Liquid
- Gene ID:101087830, 10989, 312444, 76614, 475764
- Antigen synonyms:Mitochondria-associated protein
- Storage buffer:whole serum, 0.05% sodium azide
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Shipping temperature:Ice
- Immunogen:Recombinant human Mitofilin.
- Size:100 µL
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Mitofilin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: