Anti-SERPIND1 Sheep Polyclonal Antibody
PIEAPA143055
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade D (heparin cofactor), member 1
- Antigen symbol:SERPIND1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:3053
- Antigen synonyms:Heparin Cofactor II
- Storage buffer:50% water, 50% glycerol, no preservative
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Shipping temperature:Ice
- Immunogen:Human Heparin Cofactor II.
- Purification:Ammonium sulfate precipitation, Ion-exchange chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: SERPIND1 (serpin peptidase inhibitor, clade D (heparin cofactor), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human