Anti-MORF4L2 Goat Polyclonal Antibody
PIEAPA131240
- Antibody type:Primary
- Antigen name:mortality factor 4 like 2
- Antigen symbol:MORF4L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog
- Western blot:Yes
- Form:Liquid
- Gene ID:481001, 9643
- Antigen synonyms:MRGX|KIAA0026|RP5-1055C14.2|MORFL2
- Storage buffer:TBS, 0.5% BSA, 0.02% sodium azide, pH 7.3
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:synthetic peptide SSRKQGSQPRGQQS (Human) (N terminal).
- Purification:Antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: MORF4L2 (mortality factor 4 like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human