- Antibody type:Primary
- Antigen name:Vomeronasal 1 receptor 1
- Antigen symbol:VN1R1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Format:whole serum
- Form:Lyophilized
- Gene ID:57191
- Antigen synonyms:V3r-related gene protein|V1r-like receptor 1|Vomeronasal olfactory receptor chromosome 19 subtype I member 1|Vomeronasal type-1 receptor 1|hGPCR24
- Storage buffer:whole serum, no preservative
- Storage temperature:Store at 4 °C short term. For long term storage, store at –20 °C, avoiding freeze/thaw cycles.
- Shipping temperature:Ice
- Immunogen:A synthetic peptide from human vomeronasal 1 receptor 1
- Size:100 µL
- Pk:100 µl
Reconstitute with 100 ul of distilled water. Centrifuge to remove any insoluble material. Pheromones are chemical signals that elicit specific behavioral responses and physiologic alterations in recipients of the same species. The protein encoded by this gene is similar to pheromone receptors and is primarily localized to the olfactory mucosa. An alternate splice variant of this gene is thought to exist, but its full length nature has not been determined. Putative pheromone receptor. Expressed in the olfactory mucosa, very low expression in brain, lung and kidney. The chimpanzee and orangutan orthologous proteins do not exist, their genes are pseudogenes.
Type: Primary
Antigen: VN1R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human