- Antibody type:Primary
- Antigen name:Glyceraldehyde-3-phosphate dehydrogenase
- Antigen symbol:GAPDH
- Clonality:Monoclonal
- Clone:GA1R
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Yeast,Hamster,Rabbit,Bacteria,Insect,Chicken,Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:374193, 24383, 100042025, 100009074, 2597
- Antigen synonyms:GAPDH|G3PD|CDABP0047|GAPD|Glyceraldehyde-3-phosphate dehydrogenase|Peptidyl-cysteine S-nitrosylase GAPDH|HEL-S-162eP|OK/SW-cl.12
- Storage buffer:PBS, no preservative, pH 7.2
- Storage temperature:Store at 4 °C short term. For long term storage, store at –20 °C, avoiding freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant GAPDH
- Purification:Protein A
- Size:100 µg
- Pk:100 µG
MA5-15738 detects GAPDH from BL-21 bacteria, Sf9 insect, Saccharomyces cerevisiae (yeast), human, mouse, rat, rabbit, hamster, and chicken samples. MA5-15738 has been successfully used in Western blot, ICC, IF, IHC (P), FACS and ELISA. MA5-15738 may be stored at 4ºC short term. Add 0.05% sodium azide if desired. For long term storage, store at -20ºC, avoiding freeze/thaw cycles. Glyceraldehyde 3 phosphate dehydrogenase (GAPDH) is an enzyme involved in glycolysis. In addition to this role, it is involved in other cellular processes ranging from membrane fusion, neuronal apoptosis to cancer. GAPDH is expressed at high levels in most tissues and is therefore useful as protein loading control in western blot analysis. Additionally, the antigen is conserved in most eukaryotes.
Type: Primary
Antigen: GAPDH
Clonality: Monoclonal
Clone: GA1R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Hamster, Bacteria, Chicken, Rabbit, Insect, Rat, Mouse, Yeast