Anti-RFP Tag Mouse Monoclonal Antibody [clone: RF5R]
PIEAMA515257
- Antibody type:Primary
- Antigen symbol:RFP Tag
- Clonality:Monoclonal
- Clone:RF5R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:RF5R
- Storage buffer:PBS, no preservative, pH 7.2
- Storage temperature:Store at 4 °C short term. For long term storage, store at –20 °C, avoiding freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:RFP from the Discosoma sea anemone N-terminal peptide-KLH conjugated.
- Purification:Protein A
- Size:100 µg
- Pk:100 µG
MA5-15257 recognizes native and denatured forms of RFP (Red Fluorescent Protein) and its many variants: tag-RFP, turbo-RFP, DsRed, mCherry, mOrange, etc. MA5-15257 has successfully been used in immunoprecipitation, ELISA, Western blotting, immunohistochemistry, immunocytochemistry and immunofluorescence applications. MA5-15257 may be stored at 4ºC short term. Add 0.05% sodium azide if desired. For long term storage, store at -20ºC, avoiding freeze/thaw cycles."
Type: Primary
Antigen: RFP Tag
Clonality: Monoclonal
Clone: RF5R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Tag