Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 7
- Antigen symbol:MCP3
- Clonality:Monoclonal
- Clone:36320.11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:6354
- Antigen synonyms:MCP3|NC28|MGC138465|SCYA7|SCYA6|MCP-3|MARC|MGC138463|FIC
- Storage buffer:PBS, no preservative, pH 7.4
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant full length protein (Human).
- Purification:Protein G
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MA1-91619 detects MCP3 from human samples. MA1-91619 has been successfully used in Western blot applications. The MA1-91619 immunogen is: Recombinant full length protein (Human). This gene encodes monocyte chemotactic protein 3, a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix. This gene is part of a cluster of C-C chemokine family members on chromosome 17q.
Type: Primary
Antigen: MCP3
Clonality: Monoclonal
Clone: 36320.11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human