Anti-PTPRB Mouse Monoclonal Antibody [clone: 122.2]
PIEAMA15870
- Antibody type:Primary
- Antigen name:Tyrosine Phosphatase beta Receptor
- Antigen symbol:PTPRB
- Clonality:Monoclonal
- Clone:122.2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:314843
- Antigen synonyms:Ptpz
- Storage buffer:PBS, no preservative
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Shipping temperature:Ice
- Immunogen:Embryonic rat brain proteoglycans.
- Purification:Size-exclusion - Dialysis
- Size:100 µL
- Pk:100 µl
MA1-5870 detects Tyrosine Phosphatase Beta Receptor in rat samples. MA1-5870 has been successfully used in Western blot, immunofluorescence, immunocytochemistry and immunoprecipitation procedures. The MA1-5870 immunogen is embryonic rat brain proteoglycans. alternative splicing product phosphocan inhibits neural cell adhesion and neurite outgrowth.
Type: Primary
Antigen: PTPRB
Clonality: Monoclonal
Clone: 122.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Rat