Specifications
- Antibody type:Primary
- Antigen name:CD1a molecule
- Antigen symbol:CD1A
- Clonality:Monoclonal
- Clone:CBT6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:909, 111334
- Antigen synonyms:FCB6|cortical thymocyte antigen CD1A|T-cell surface antigen T6/Leu-6|cluster of differentiation 1 A|T6|CD1|R4|a polypeptide|T-cell surface glycoprotein CD1a|CD1A antigen|HTA1
- Storage buffer:PBS, 0.05% sodium azide
- Storage temperature:4° C
- Concentration:0.1 mg/ml
- Shipping temperature:Ice
- Immunogen:Human CD1a.
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: CBT6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human