Anti-CTSL Mouse Monoclonal Antibody [clone: 33/2]
PIEAMA126774
- Antibody type:Primary
- Antigen name:Cathepsin L
- Antigen symbol:CTSL
- Clonality:Monoclonal
- Clone:33/2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Mustelid,Human,Rat,Mouse
- Western blot:Yes
- Format:Ascites
- Form:Liquid
- Gene ID:1514, 25697, 13039
- Antigen synonyms:nkt|Ctsl1|MEP|1190035F06Rik|fs
- Storage buffer:Ascites, 15mM sodium azide
- Storage temperature:Store at 4 °C short term. For long term storage, store at –20 °C, avoiding freeze/thaw cycles.
- Shipping temperature:Ice
- Immunogen:Purified, full length, native protein from human lung cancer line EPLC 32 M1.
- Size:100 µL
- Pk:100 µl
MA1-26774 detects Cathepsin L from human, mouse (weakly), rat, and mink samples. MA1-26774 has been successfully used in immunohistochemistry (frozen), ELISA, and Western blot procedures. The MA1-26774 immunogen is purified, full length, native protein from human lung cancer line EPLC 32 M1. Store at 4ºC short term. For extended storage, aliquot and store at -20ºC or below. Avoid freeze/thaw cycles.
Type: Primary
Antigen: CTSL
Clonality: Monoclonal
Clone: 33/2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mustelid, Mouse