Anti-TIRAP Mouse Polyclonal Antibody
ABNOH00114609-B01
: Abnova
- Antibody type:Primary
- Antigen name:Toll-interleukin 1 receptor (TIR) domain containing adaptor protein
- Antigen symbol:TIRAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:114609
- Antigen synonyms:TIRAP|MyD88 adapter-like protein|TIR domain-containing adapter protein|MyD88-2|Toll/interleukin-1 receptor domain-containing adapter protein|MAL|Adaptor protein Wyatt
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: TIRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human