Anti-IMMP2 Mouse Polyclonal Antibody
50 µl
Catalog # ABNOH00083943-A01
Jump to:
Discontinued
Product Details & Documents
Type: Primary
Antigen: IMMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Documents
TDS_ABNOH00083943-A01.pdf
Datasheet
Specifications
- Size:50 µl
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Mouse
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:Inner Mitochondrial Membrane Peptidase Subunit 2
- Cat. No.:ABNOH00083943-A01
- Reactivity:Human
- Gene ID:83943
- Antigen synonyms:IMMP2L-IT1|IMP2|IMP2-LIKE
- Conjugation:Unconjugated
- Antigen symbol:IMMP2
- ELISA:Yes



