Anti-MIXL1 Mouse Monoclonal Antibody [clone: 4D11]
ABNOH00083881-M02
: Abnova
- Antibody type:Primary
- Antigen name:Mix paired-like homeobox
- Antigen symbol:MIXL1
- Clonality:Monoclonal
- Clone:4D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:83881
- Antigen synonyms:MIXL|MILD1|MIX
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MIXL1
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human