Anti-ILKAP Mouse Polyclonal Antibody
Catalog # ABNOH00080895-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Integrin-Linked Kinase-Associated Serine/Threonine Phosphatase
- Antigen symbol:ILKAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:80895
- Antigen synonyms:ILKAP3|ILKAP2|PP2C-DELTA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ILKAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human