Anti-CDG1G, ECM39, PP14673, hALG12 Mouse Monoclonal Antibody [clone: 5E3]
Catalog # ABNOH00079087-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Alpha-1,6-MannosyltransferaseAsparagine-Linked Glycosylation 12
- Antigen symbol:CDG1G, ECM39, PP14673, hALG12
- Clonality:Monoclonal
- Clone:5E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79087
- Antigen synonyms:hALG12|ECM39|CDG1G|PP14673
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CDG1G, ECM39, PP14673, hALG12
Clonality: Monoclonal
Clone: 5E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat