Anti-KLHL1 Mouse Monoclonal Antibody [clone: 3C2]
Catalog # ABNOH00057626-M09
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Kelch-like 1
- Antigen symbol:KLHL1
- Clonality:Monoclonal
- Clone:3C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:57626
- Antigen synonyms:MRP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: KLHL1
Clonality: Monoclonal
Clone: 3C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human