Anti-TTYH1 Mouse Monoclonal Antibody [clone: 4A9]
ABNOH00057348-M04
: Abnova
- Antibody type:Primary
- Antigen name:Tweety Family Member 1
- Antigen symbol:TTYH1
- Clonality:Monoclonal
- Clone:4A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:57348
- Antigen synonyms:TTYH1|Protein tweety homolog 1|hTTY1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TTYH1
Clonality: Monoclonal
Clone: 4A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human