Anti-MTFP1 Mouse Polyclonal Antibody
Catalog # ABNOH00051537-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mitochondrial Fission Process 1
- Antigen symbol:MTFP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51537
- Antigen synonyms:MTP18|HSPC242
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: MTFP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human