Anti-QPCT Rabbit Polyclonal Antibody
ABNOH00025797-D01
: Abnova
- Antibody type:Primary
- Antigen name:Glutaminyl-peptide cyclotransferase
- Antigen symbol:QPCT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:25797
- Antigen synonyms:QC|sQC|GCT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: QPCT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human