Anti-RAD54B Rabbit Polyclonal Antibody
Catalog # ABNOH00025788-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:RAD54 homolog B
- Antigen symbol:RAD54B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:25788
- Antigen synonyms:DNA repair and recombination protein RAD54B|Fibrinogen silencer binding protein|RAD54 homolog B|RA54B_HUMAN.|FSBP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: RAD54B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human