Anti-RASGRP3 Rabbit Polyclonal Antibody
ABNOH00025780-D01P
: Abnova
- Antibody type:Primary
- Antigen name:RAS guanyl releasing protein 3 (calcium and DAG-regulated)
- Antigen symbol:RASGRP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:25780
- Antigen synonyms:GRP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RASGRP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse