- Antibody type:Primary
- Antigen name:Leucine zipper protein down-regulated in cancer cells
- Antigen symbol:LDOC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:23641
- Antigen synonyms:Leucine zipper protein down-regulated in cancer cells|Protein LDOC1|LDOC1|Mar7|BCUR1|Mart7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: LDOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human