Anti-WDFY3 Mouse Monoclonal Antibody [clone: 2F12]
Catalog # ABNOH00023001-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:WD Repeat And FYVE Domain Containing 3
- Antigen symbol:WDFY3
- Clonality:Monoclonal
- Clone:2F12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:23001
- Antigen synonyms:ALFY|ZFYVE25
- Storage buffer:In 1X PBS, pH 7,4
- Sequence:MGTSKEKAKTATLKQALLGHTDTVTCATASLAYHIIVSGSRDRTCIIWDLNKLSFLTQLRGHRAPVSALCINELTGDIVSCAGTYIHVWSINGNPIVSVNTFTGRSQQIICCCMSEMNEWDTQNVIVTGHSDGVVRFWRMEFLQVPETPAPEPAEVLEMQEDCPEAQIGQEAQDEDSSDSEADEQSISQDPKDTPSQPSSTSHRPRAASCRATAAWCTDSGSDDSRRWSDQLSLDEKDGFIFVNYSEGQTRAHLQGPLSHPHPNPIEVRNYSRLKPGYRWERQLVFRSKLTMHTAFDRKDNAHPAEVTALGISKDHSRILVGDSRGRVFSWSVSDQPGRSAADHWVKDEGGDSCSGCSVRFSLTERRHHCRNCGQLFCQKCSRFQSEIKRLKISSPVRVCQNCYYNLQHERGSEDGPRNC
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:WDFY3 (AAH15214, 1 a.a. ~ 420 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 kDa.
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a full-length recombinant WDFY3.
Recommended Dilutons: Western Blot (1 to 5 µg/ml).
Type: Primary
Antigen: WDFY3
Clonality: Monoclonal
Clone: 2F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human