Anti-ECD Mouse Monoclonal Antibody [clone: 1A11]
Catalog # ABNOH00011319-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ecdysoneless homolog
- Antigen symbol:ECD
- Clonality:Monoclonal
- Clone:1A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11319
- Antigen synonyms:GCR2|HSGT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ECD
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human