Anti-RNF41 Mouse Monoclonal Antibody [clone: 4C2]
ABNOH00010193-M01A
: Abnova
- Antibody type:Primary
- Antigen name:Ring Finger Protein 41
- Antigen symbol:RNF41
- Clonality:Monoclonal
- Clone:4C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10193
- Antigen synonyms:FLRF|SBBI03|NRDP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: RNF41
Clonality: Monoclonal
Clone: 4C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat