Anti-EDIL3 Mouse Polyclonal Antibody
Catalog # ABNOH00010085-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:EGF-Like Repeats And Discoidin I-Like Domains 3
- Antigen symbol:EDIL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10085
- Antigen synonyms:Developmental Endothelial Locus-1|Integrin-Binding Protein DEL1|DEL1|Developmentally-Regulated Endothelial Cell Locus 1 Protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: EDIL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human