Anti-USH1C Mouse Monoclonal Antibody [clone: 2B9]
Catalog # ABNOH00010083-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Usher syndrome 1C
- Antigen symbol:USH1C
- Clonality:Monoclonal
- Clone:2B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10083
- Antigen synonyms:AIE-75|PDZ-73/NY-CO-38|deafness|HARMONIN|Usher syndrome 1C (autosomal recessive|NY-CO-38|severe)|PDZ-73|DFNB18|PDZ-73 protein|USH1C|autosomal recessive 18|harmonin|NY-CO-37|ush1cpst|PDZ-45
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: USH1C
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human