Anti-TSPAN32 Mouse Monoclonal Antibody [clone: 2G12]
ABNOH00010077-M04
: Abnova
- Antibody type:Primary
- Antigen name:Tetraspanin 32
- Antigen symbol:TSPAN32
- Clonality:Monoclonal
- Clone:2G12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10077
- Antigen synonyms:ART1, PHEMX, PHMX, TSSC6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TSPAN32
Clonality: Monoclonal
Clone: 2G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human