Anti-DPP3 Mouse Monoclonal Antibody [clone: 4D3]
Catalog # ABNOH00010072-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Dipeptidylpeptidase 3
- Antigen symbol:DPP3
- Clonality:Monoclonal
- Clone:4D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10072
- Antigen synonyms:DPPIII
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: DPP3
Clonality: Monoclonal
Clone: 4D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human